5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide ≥98%

Supplier: Matrix Scientific

Synonyms: 5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulfonamidePazopanib5-((4-((2,3-Dimethyl-2H-indazol-6-yl)(methyl)amino)pyrimidin-2-yl)amino)-2-methylbenzenesulphonamide

068699-1G
101937-294EA 1073.85 USD
101937-294
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide ≥98%
5-[4-[(2,3-Dimethyl-2H-indazol-6-yl)methylamino]pyrimidin-2-ylamino]-2-methylbenzenesulfonamide

Hazard: Irritant

Formula: C₂₁H₂₃N₇O₂S
MW: 437.53 g/mol
MDL Number: MFCD11616589
CAS Number: 444731-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR