5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥95%

Supplier: Matrix Scientific

Synonyms: 5-bromo-4-chloro-2-(methylsulfanyl)pyrimidine5-bromo-4-chloro-2-(methylsulphanyl)pyrimidine

068693-1G
101937-266EA 36.92 USD
101937-266
5-Bromo-4-chloro-2-(methylthio)pyrimidine ≥95%
5-Bromo-4-chloro-2-(methylthio)pyrimidine

Hazard: Irritant

Formula: C₅H₄BrClN₂S
MW: 239.5200 g/mol
MDL Number: MFCD00023232
CAS Number: 63810-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR