Imidazo[1,2-a]pyridine-8-carboxylic acid ≥95%

Supplier: Matrix Scientific

068682-1G
101937-220EA 673.9 USD
101937-220
Imidazo[1,2-a]pyridine-8-carboxylic acid ≥95%
Imidazo[1,2-a]pyridine-8-carboxylic acid

Hazard: Irritant

Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
MDL Number: MFCD06739251
CAS Number: 133427-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR