2-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-Isoindole-1,3(2H)-dione ≥95%

Supplier: Matrix Scientific

068678-250MG 068678-1G
101937-200EA 407.69 USD
101937-200 101937-204
2-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-Isoindole-1,3(2H)-dione ≥95%
2-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)-1H-Isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₄H₈N₄O₂
MW: 264.25 g/mol
MDL Number: MFCD11045038
CAS Number: 741686-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR