(S)-Tetrahydrofuran-3-ylmethanol ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-(Tetrahydrofuran-3-yl)methanol

068673-500MG
101937-184EA 720 USD
101937-184
(S)-Tetrahydrofuran-3-ylmethanol ≥95%
(S)-Tetrahydrofuran-3-ylmethanol

Hazard: Irritant

Formula: C₅H₁₀O₂
MW: 102.13 g/mol
MDL Number: MFCD17015961
CAS Number: 124391-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR