(±)-Tetrahydrofuran-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Tetrahydro-3-furoic acidTetrahydrofuran-3-carboxylic acid

068671-1G
101937-176EA 15.38 USD
101937-176
(±)-Tetrahydrofuran-3-carboxylic acid ≥95%
(±)-Tetrahydro-2-furoic acid

Hazard: Irritant/Refrigerate

Formula: C₅H₈O₃
MW: 116.1152 g/mol
Boiling Pt: 140…141 °C (15 mmHg)
Density: 1.21
Flash Pt: 113 °C
MDL Number: MFCD00075173
CAS Number: 89364-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR