4,5-Dichloro-2-(methylsulfanyl)pyrimidine ≥95%
Supplier: Matrix Scientific
Synonyms:
4,5-Dichloro-2-(methylthio)pyrimidine
068661-5G
101937-138EA
426.15
USD
101937-138
4,5-Dichloro-2-(methylsulfanyl)pyrimidine ≥95%
4,5-Dichloro-2-(methylsulfanyl)pyrimidine
Hazard: Irritant
Formula:
C₅H₄Cl₂N₂S MW: 195.07 g/mol |
MDL Number:
MFCD16987934 CAS Number: 99469-85-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...