1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Cyano-7-azaindole
068657-250MG
068657-1G
068657-500MG
101937-118EA
1133.85
USD
101937-118
101937-122
101937-120
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile ≥95%
1H-Pyrrolo[2,3-b]pyridine-6-carbonitrile
Hazard: Irritant
Formula:
C₈H₅N₃ MW: 143.15 g/mol |
MDL Number:
MFCD06659673 CAS Number: 189882-33-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...