3,3-Difluorocyclobutanamine hydrochloride ≥95%

Supplier: Matrix Scientific

068653-1G
101937-102EA 75.38 USD
101937-102
3,3-Difluorocyclobutanamine hydrochloride ≥95%
3,3-Difluorocyclobutanamine hydrochloride

Hazard: Irritant

Formula: C₄H₈ClF₂N
Melting Pt: >300 °C
MDL Number: MFCD10001382
CAS Number: 637031-93-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR