3-Aminocyclobutanol hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Aminocyclobutanol

068650-1G
101937-084EA 230.77 USD
101937-084
3-Aminocyclobutanol hydrochloride ≥95%
3-Aminocyclobutanol hydrochloride

Hazard: Irritant

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD12547215
CAS Number: 4640-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR