3-Fluorocyclobutanecarboxylic acid ≥95%
Supplier: Matrix Scientific
068648-1G
068648-500MG
068648-250MG
101937-068EA
669.23
USD
101937-068
101937-070
101937-072
3-Fluorocyclobutanecarboxylic acid ≥95%
3-Fluorocyclobutanecarboxylic acid
Hazard: Irritant
Formula:
C₅H₇FO₂ |
MDL Number:
MFCD09834117 CAS Number: 122665-96-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...