Methyl-3,3-difluorocyclobutanecarboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Methyl 3,3-difluorocyclobutanecarboxylate
, Methyl 3,3-difluorocyclobutane-1-carboxylate
068646-500MG
068646-250MG
101937-058EA
466.15
USD
101937-058
101937-060
Methyl-3,3-difluorocyclobutanecarboxylate ≥95%
Methyl-3,3-difluorocyclobutanecarboxylate
Hazard: Irritant
Formula:
C₆H₈F₂O₂ MW: 150.13 g/mol |
MDL Number:
MFCD17015922 CAS Number: 1234616-13-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...