tert-Butyl-3-hydroxycyclobutylcarbamate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl (3-hydroxycyclobutyl)carbamatetert-butyl N-(3-hydroxycyclobutyl)carbamate

068644-250MG
101937-048EA 152.31 USD
101937-048
tert-Butyl-3-hydroxycyclobutylcarbamate ≥95%
tert-Butyl-3-hydroxycyclobutylcarbamate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
MDL Number: MFCD11501030
CAS Number: 154748-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR