(S)-3-(Methylsulfonyl)pyrrolidine ≥95%

Supplier: Matrix Scientific

068626-250MG 068626-500MG
101936-978EA 261.54 USD
101936-978 101936-980
(S)-3-(Methylsulfonyl)pyrrolidine ≥95%
(S)-3-(Methylsulfonyl)pyrrolidine

Hazard: Irritant

Formula: C₅H₁₁NO₂S
MW: 149.21 g/mol
MDL Number: MFCD16294584
CAS Number: 290328-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR