3-Hydroxypiperidine hydrochloride ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Hydroxypiperidine Hydrochloride
068616-25G
068616-1G
101936-932EA
1252.03
USD
101936-932
101936-936
3-Hydroxypiperidine hydrochloride ≥95%
3-Hydroxypiperidine hydrochloride
Hazard: Irritant
Formula:
C₅H₁₂ClNO MW: 137.61 g/mol |
MDL Number:
MFCD00012772 CAS Number: 64051-79-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...