Imidazo[1,2-b]pyridazine-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
068612-250MG
068612-500MG
101936-912EA
163.3
USD
101936-912
101936-914
Imidazo[1,2-b]pyridazine-6-carboxylic acid ≥95%
Imidazo[1,2-b]pyridazine-6-carboxylic acid
Hazard: Irritant
Formula:
C₇H₅N₃O₂ MW: 163.1300 g/mol |
MDL Number:
MFCD11044772 CAS Number: 769109-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...