3-Amino-6-bromopyridazine ≥95%
Supplier: Matrix Scientific
Synonyms:
6-Bromopyridazin-3-amine
068608-1G
068608-5G
101936-898EA
376.92
USD
101936-898
101936-896
3-Amino-6-bromopyridazine ≥95%
3-Amino-6-bromopyridazine
Hazard: Irritant
Formula:
C₄H₄BrN₃ MW: 173.9987 g/mol |
MDL Number:
MFCD01646092 CAS Number: 88497-27-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...