3-Pyrrolidineacetic acid ≥95%
Supplier: Matrix Scientific
068600-1G
068600-5G
101936-862EA
2692.31
USD
101936-862
101936-860
3-Pyrrolidineacetic acid ≥95%
3-Pyrrolidineacetic acid
Hazard: Irritant
Formula:
C₁₂H₂₁NO₄ |
MDL Number:
MFCD18910910 CAS Number: 890849-27-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...