trans-3-(Boc-amino)cyclobutanecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
(1r,3r)-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid
, trans-3-((tert-Butoxycarbonyl)amino)cyclobutanecarboxylic acid
068591-500MG
068591-250MG
101936-818EA
224.62
USD
101936-818
101936-816
trans-3-(Boc-amino)cyclobutanecarboxylic acid ≥95%
trans-3-(Boc-amino)cyclobutanecarboxylic acid
Hazard: Irritant
Formula:
C₁₀H₁₇NO₄ MW: 215.249 g/mol |
MDL Number:
MFCD06657680 CAS Number: 939400-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...