tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥96%

Supplier: Matrix Scientific

Synonyms: 1-BOC-7-Azaindole-3-boronic acid, pinacol ester

068576-1G
101936-750EA 630.77 USD
101936-750
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate ≥96%
tert-Butyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine-1-carboxylate

Hazard: Irritant

Formula: C₁₈H₂₅BN₂O₄
MW: 344.22 g/mol
MDL Number: MFCD12407268
CAS Number: 942070-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR