4-(Aminomethyl)-1-methylpiperidin-2-one ≥95%

Supplier: Matrix Scientific

068573-250MG 068573-500MG
101936-734EA 492.31 USD
101936-734 101936-736
4-(Aminomethyl)-1-methylpiperidin-2-one ≥95%
4-(Aminomethyl)-1-methylpiperidin-2-one

Hazard: Irritant

Formula: C₇H₁₄N₂O
MDL Number: MFCD17015880
CAS Number: 1234616-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR