Methyl-3-oxocyclobutanecarboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-oxocyclobutanecarboxylate

068561-5G
101936-688EA 30.77 USD
101936-688
Methyl-3-oxocyclobutanecarboxylate ≥95%
Methyl-3-oxocyclobutanecarboxylate

Hazard: Irritant

Formula: C₆H₈O₃
MW: 128.127 g/mol
MDL Number: MFCD00604222
CAS Number: 695-95-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR