1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine ≥95%
Supplier: Matrix Scientific
068489-1G
068489-5G
101936-370EA
2733.85
USD
101936-370
101936-372
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine ≥95%
1-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2,3,6-tetrahydropyridine
Hazard: Irritant
Formula:
C₁₂H₂₂BNO₂ |
MDL Number:
MFCD16039627 CAS Number: 1254982-25-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...