2-Methylindazole-6-carboxylic acid ≥95%
Supplier: Matrix Scientific
068466-250MG
068466-1G
068466-500MG
101936-274EA
660
USD
101936-274
101936-276
101936-272
2-Methylindazole-6-carboxylic acid ≥95%
2-Methylindazole-6-carboxylic acid
Hazard: Irritant
Formula:
C₉H₈N₂O₂ |
MDL Number:
MFCD15071449 CAS Number: 1031417-46-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...