2-Amino-5-pyrimidineboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Aminopyrimidine-5-boronic acid(2-Aminopyrimidin-5-yl)boronic acid

068426-1G 068426-5G
101936-084EA 267.69 USD
101936-084 101936-086
2-Amino-5-pyrimidineboronic acid ≥95%
2-Amino-5-pyrimidineboronic acid

Hazard: Irritant

Formula: C₄H₆BN₃O₂
MW: 138.92 g/mol
MDL Number: MFCD07375147
CAS Number: 936250-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR