1-Boc-4-amino-4-methylpiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 4-amino-4-methylpiperidine-1-carboxylate1-Boc-4-Amino-4-methylpiperidine

068415-250MG
101936-038EA 53.85 USD
101936-038
1-Boc-4-amino-4-methylpiperidine ≥95%
1-Boc-4-amino-4-methylpiperidine

Hazard: Irritant

Formula: C₁₁H₂₂N₂O₂
MW: 214.3100 g/mol
MDL Number: MFCD03265497
CAS Number: 343788-69-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR