3-(3-Pyrazin-2-ylpropyl)azetidine-3-carboxylic acid dimethylamide

Supplier: Matrix Scientific

068372-250MG
101935-862EA 638.46 USD
101935-862
3-(3-Pyrazin-2-ylpropyl)azetidine-3-carboxylic acid dimethylamide
3-(3-Pyrazin-2-ylpropyl)azetidine-3-carboxylic acid dimethylamide

Hazard: Irritant

Formula: C₁₃H₂₀N₄O
MDL Number: MFCD21606283
CAS Number: 1361112-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR