1-Pyridazin-3-ylpyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

068304-250MG
101935-726EA 558.46 USD
101935-726
1-Pyridazin-3-ylpyrrolidine-3-carboxylic acid
1-Pyridazin-3-ylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₁N₃O₂
MDL Number: MFCD17272788
CAS Number: 1316219-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR