Methyl(6-methyl-2-piperidin-4-ylmethylpyrimidin-4-yl)amine dihydrochloride

Supplier: Matrix Scientific

068276-250MG
101935-670EA 687.69 USD
101935-670
Methyl(6-methyl-2-piperidin-4-ylmethylpyrimidin-4-yl)amine dihydrochloride
Methyl(6-methyl-2-piperidin-4-ylmethylpyrimidin-4-yl)amine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₂Cl₂N₄
MDL Number: MFCD21606208
CAS Number: 1361116-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR