3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
Supplier: Matrix Scientific
Synonyms:
3-Fluoro-5-(trifluoromethyl)picolinonitrile
067180-500MG
067180-1G
101933-416EA
346.15
USD
101933-416
101933-414
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
3-Fluoro-5-(trifluoromethyl)pyridine-2-carbonitrile
Hazard: Irritant
Formula:
C₇H₂F₄N₂ MW: 190.1 g/mol |
MDL Number:
MFCD11848039 CAS Number: 80194-71-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...