6-Bromo-2-(methylamino)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-N-methyl-pyridin-2-amine6-Bromo-N-methylpyridin-2-amine

067110-1G
101933-182EA 85.1 USD
101933-182
6-Bromo-2-(methylamino)pyridine ≥95%
6-Bromo-2-(methylamino)pyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MW: 187.0400 g/mol
MDL Number: MFCD09261221
CAS Number: 89026-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR