(1-Bromo-2,2,2-trifluoroethyl)benzene ≥95%

Supplier: Matrix Scientific

067091-1G
101933-164EA 144.62 USD
101933-164
(1-Bromo-2,2,2-trifluoroethyl)benzene ≥95%
(1-Bromo-2,2,2-trifluoroethyl)benzene

Hazard: Irritant

Formula: C₈H₆BrF₃
MDL Number: MFCD12789673
CAS Number: 434-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR