Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Benzhydryl-2-azaspiro[3.3]heptan-5-ol ≥95%

Supplier: Matrix Scientific

067055-250MG
101933-080
2-Benzhydryl-2-azaspiro[3.3]heptan-5-ol ≥95%
Purity: 95%
CAS: 1263296-81-6
MDL Number: MFCD20444452
Molecular Formula: C19H21NO
Molecular Weight: 279,39
Hazard: Irritant


Formula: C₁₉H₂₁NO
MW: 279,39 g/mol
MDL Number: MFCD20444452
CAS Number: 1263296-81-6
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty