MATR067038

067038-250MG
101933-042EA 810.77 USD
101933-042
MATR067038
Purity: 95%
CAS: 933727-35-6
MDL Number: MFCD20444448
Molecular Formula: C6H10O3
Molecular Weight: 130,14
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR