3-(Nitromethylene)oxetane ≥95%

Supplier: Matrix Scientific

067035-1G
101933-034EA 1192.31 USD
101933-034
3-(Nitromethylene)oxetane ≥95%
3-(Nitromethylene)oxetane

Hazard: Irritant/Store in Freezer

Formula: C₄H₅NO₃
MDL Number: MFCD12547195
CAS Number: 922500-95-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR