2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile ≥95%
Supplier: Matrix Scientific
066926-1G
066926-5G
066926-500MG
101932-774EA
403.08
USD
101932-774
101932-776
101932-778
2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile ≥95%
2-[Methoxy(naphthalen-1-yl)methylidene]propanedinitrile
Hazard: Irritant
Formula:
C₁₅H₁₀N₂O MW: 234.26 g/mol |
MDL Number:
MFCD04116187 CAS Number: 221242-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...