1-(Chloromethyl)-2-fluoro-4-nitrobenzene ≥95%
Supplier: Matrix Scientific
066875-1G
066875-500MG
101932-588EA
803.08
USD
101932-588
101932-590
1-(Chloromethyl)-2-fluoro-4-nitrobenzene ≥95%
1-(Chloromethyl)-2-fluoro-4-nitrobenzene
Hazard: Irritant
Formula:
C₇H₅ClFNO₂ |
MDL Number:
MFCD11110171 CAS Number: 1159607-50-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...