6-Chloro-2-hydroxy-1,8-naphthyridine-3-carbonitrile ≥95%

Supplier: Matrix Scientific

066871-500MG 066871-1G
101932-572EA 803.08 USD
101932-572 101932-574
6-Chloro-2-hydroxy-1,8-naphthyridine-3-carbonitrile ≥95%
6-Chloro-2-hydroxy-1,8-naphthyridine-3-carbonitrile

Hazard: Irritant

Formula: C₉H₄ClN₃O
MDL Number: MFCD19982796
CAS Number: 1330583-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR