4-Cyanopyridine-N-oxide ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Cyanopyridine 1-oxide

066833-1G
101932-498EA 138.46 USD
101932-498
4-Cyanopyridine-N-oxide ≥98%
4-Cyanopyridine N-oxide

Hazard: Irritant

Formula: C₆H₄N₂O
MW: 120.1088 g/mol
MDL Number: MFCD00006205
CAS Number: 14906-59-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR