4-(Prop-2-yn-1-yloxy)but-1-yne ≥95%

Supplier: Matrix Scientific

066816-1G
101932-464EA 364.62 USD
101932-464
4-(Prop-2-yn-1-yloxy)but-1-yne ≥95%
4-(Prop-2-yn-1-yloxy)but-1-yne

Hazard: Irritant

Formula: C₇H₈O
MDL Number: MFCD19982828
CAS Number: 75405-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR