Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Supplier: Matrix Scientific
066784-1G
066784-500MG
101932-334EA
600
USD
101932-334
101932-336
Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate ≥95%
Benzyl-4-{[(3-fluorophenyl)amino]methyl}-4-hydroxypiperidine-1-carboxylate
Hazard: Irritant
Formula:
C₂₀H₂₃FN₂O₃ MW: 358.42 g/mol |
MDL Number:
MFCD19686118 CAS Number: 1047655-89-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...