5-Hydrazinyl-2-(trifluoromethyl)pyridine

Supplier: Matrix Scientific

066775-1G
101932-312EA 190.77 USD
101932-312
5-Hydrazinyl-2-(trifluoromethyl)pyridine
5-Hydrazinyl-2-(trifluoromethyl)pyridine

Hazard: Toxic

Formula: C₆H₆F₃N₃
MDL Number: MFCD11113388
CAS Number: 1035173-53-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR