tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate

Supplier: Matrix Scientific

Synonyms: 2-Oxo-1,9-diazaspiro[5.5]undecane, N9-BOC protected

066759-250MG
101932-282EA 444.62 USD
101932-282
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate
tert-Butyl-2-oxo-1,9-diazaspiro[5.5]undecane-9-carboxylate

Hazard: Irritant

Formula: C₁₄H₂₄N₂O₃
MW: 268.36 g/mol
MDL Number: MFCD09998742
CAS Number: 1031927-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR