2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester
Supplier: Matrix Scientific
Synonyms:
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene, N9-BOC protected
066758-250MG
101932-280EA
606.15
USD
101932-280
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester
2-Oxo-1,9-diazaspiro[5.5]undec-3-ene-9-carboxylic acid tert-butyl ester
Hazard: Irritant
Formula:
C₁₄H₂₂N₂O₃ MW: 266.34 g/mol |
MDL Number:
MFCD09998740 CAS Number: 1031927-11-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...