5-Bromo-7-nitroindoline

Supplier: Matrix Scientific

066740-1G
101932-248EA 126.15 USD
101932-248
5-Bromo-7-nitroindoline
5-Bromo-7-nitroindoline

Hazard: Irritant

Formula: C₈H₇BrN₂O₂
MW: 243.06 g/mol
MDL Number: MFCD00005708
CAS Number: 80166-90-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR