3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine

Supplier: Matrix Scientific

066734-250MG
101932-236EA 343.08 USD
101932-236
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine
3-(2-Bromophenyl)-4,5,6,7-tetrahydro-2H-pyrazolo[4,3-c]pyridine

Hazard: Irritant

Formula: C₁₂H₁₂BrN₃
MDL Number: MFCD09258796
CAS Number: 916423-54-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR