5-Fluoronaphthalen-1-ol
Supplier: Matrix Scientific
066713-250MG
101932-192EA
492.31
USD
101932-192
5-Fluoronaphthalen-1-ol
5-Fluoronaphthalen-1-ol
Hazard: Irritant
Formula:
C₁₀H₇FO |
MDL Number:
MFCD12401609 CAS Number: 61761-32-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...