6-(Hydroxymethyl)picolinic acid

Supplier: Matrix Scientific

Synonyms: 6-(Hydroxymethyl)pyridine-2-carboxylic acid

066712-1G
101932-190EA 638.46 USD
101932-190
6-(Hydroxymethyl)picolinic acid
6-(Hydroxymethyl)picolinic acid

Hazard: Irritant

Formula: C₁H₁NO₃
MW: 153.1354 g/mol
MDL Number: MFCD09032021
CAS Number: 1197-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR