7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate

Supplier: Matrix Scientific

066701-1G
101932-166EA 304.62 USD
101932-166
7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate
7-Fluorobenzo[d]imidazo[2,1-b]thiazole-2-carboxylic acid hydrate

Hazard: Irritant

Formula: C₁₀H₇FN₂O₃S
MW: 254.24 g/mol
MDL Number: MFCD08445326
CAS Number: 849924-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR