2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine

Supplier: Matrix Scientific

066689-1G
101932-142EA 643.08 USD
101932-142
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine
2-(4-Acetyl-1,4-diazepan-1-yl)ethanamine

Hazard: Irritant

Formula: C₉H₁₉N₃O
MW: 185.27 g/mol
MDL Number: MFCD09258788
CAS Number: 928003-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR